Tinidazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-[2-(Ethylsulphonyl)ethyl]-2-methyl-5-nitroimidazole, 1-(2-(Ethylsulphonyl)ethyl)-2-methyl-5-nitro-1H-imidazole

A128250-1000G A128250-100G A128250-100MG A128250-1G A128250-500G A128250-25G A128250-5G
76996-196EA 215.37 USD
76996-196 76996-198 76996-200 76996-202 76996-204 77313-396 77313-398
Tinidazole 97%
Tinidazole

Precaución: for Research Use Only

Formula: C₈H₁₃N₃O₄S
MW: 247.27 g/mol
Boiling Pt: 528.4 °C (760 mm Hg)
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00057217
CAS Number: 19387-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR