Tilorone dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride, 2,7-Bis(2-(Diethylamino)ethoxy)-9H-fluoren-9-one dihydrochloride
A632643-100MG
A632643-1MG
A632643-250MG
A632643-25MG
103799-704EA
5
USD
103799-704
103840-284
76790-768
76790-770
76790-772
76790-774
Tilorone dihydrochloride 98%
Tilorone dihydrochloride
Precaución: for Research Use Only
Formula:
C₂₅H₃₄N₂O₃ MW: 483.47 g/mol Boiling Pt: 552.3 °C (760 mm Hg) Melting Pt: 230…233 °C Flash Pt: 287.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134071 CAS Number: 27591-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...