Thiophene-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde

A102004-100G A102004-10G A102004-1G A102004-500G A102004-5G A102004-25G
103821-946EA 6 USD
103821-946 76970-200 76970-202 76970-204 76970-206 77308-828 77548-924
Thiophene-3-carbaldehyde 98%
Thiophene-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 190.6 °C (760 mm Hg)
Melting Pt: …30 °C
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005466
CAS Number: 498-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR