3,4-Thiophenedicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Thiophene-3,4-dicarboxylic acid

A175960-1G A175960-5G A175960-100G A175960-10G A175960-250MG A175960-25G
103807-102EA 7 USD
103807-102 103845-414 77082-764 77082-766 77096-620 77096-622 77096-624 77301-408
3,4-Thiophenedicarboxylic acid 98%
3,4-Thiophenedicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄O₄S
Boiling Pt: 405.9 °C (760 mm Hg)
Melting Pt: 227…231 °C
Flash Pt: 199.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00067079
CAS Number: 4282-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR