Thiophene-3,4-dicarbonitrile 97%

Supplier: Ambeed
Warning

A151631-10G A151631-250MG A151631-1G A151631-5G A151631-25G
103804-286EA 20 USD
103804-286 103842-438 77095-828 77095-830 77299-890 77299-892 77318-020
Thiophene-3,4-dicarbonitrile 97%
Thiophene-3,4-dicarbonitrile

Precaución: for Research Use Only

Formula: C₆H₂N₂S
Boiling Pt: 339 °C (760 mm Hg)
Flash Pt: 158.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01233047
CAS Number: 18853-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR