2-Thiophenesulphonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: Thiophene-2-sulphonyl chloride

A258464-10G A258464-1G A258464-250MG A258464-25G A258464-5G
103852-250EA 8 USD
103852-250 77014-128 77014-130 77014-132 77091-774 77091-776
2-Thiophenesulphonyl chloride 97%
2-Thiophenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₄H₃ClO₂S₂
Melting Pt: 30…32 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005426
CAS Number: 16629-19-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR