2-Thiophenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: Thiophene-2-sulphonamide

A161544-100G A161544-10G A161544-1G A161544-25G A161544-5G
103805-500EA 9 USD
103805-500 103843-738 77286-454 77286-456 77319-720 77319-722 77319-724
2-Thiophenesulfonamide 97%
2-Thiophenesulfonamide

Precaución: for Research Use Only

Formula: C₄H₅NO₂S₂
MW: 163.21 g/mol
Boiling Pt: 338.1 °C (760 mm Hg)
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00185853
CAS Number: 6339-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR