2,5-Thiophenedicarboxylic acid 97%

Supplier: Ambeed
Warning

A115802-100G A115802-10G A115802-25G A115802-500G A115802-5G
103824-484EA 5 USD
103824-484 77284-484 77284-486 77284-488 77284-490 77311-308
2,5-Thiophenedicarboxylic acid 97%
2,5-Thiophenedicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄O₄S
Boiling Pt: 444.6 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 222.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00016896
CAS Number: 4282-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR