Thiophen-3-amine oxalate(1:x) 97%

Supplier: Ambeed
Warning

Synonyms: Thiophen-3-amine xoxalate

A130548-10G A130548-1G A130548-25G A130548-5G
103801-438EA 5 USD
103801-438 76782-316 77313-876 77313-878 77313-880
Thiophen-3-amine oxalate(1:x) 97%
Thiophen-3-amine oxalate(1:x)

Precaución: for Research Use Only

Formula: C₆H₇NO₄S
MW: 189.1891 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986878
CAS Number: 861965-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR