Thieno[3,2-b]thiophene 98%
Supplier: Ambeed
|
Warning
|
A104028-250MG
A104028-1G
A104028-25G
A104028-100G
103796-642EA
11
USD
103796-642
103822-354
77107-390
77309-216
77548-978
77614-786
Thieno[3,2-b]thiophene 98%
Thieno[3,2-b]thiophene
Precaución: for Research Use Only
Formula:
C₆H₄S₂ Melting Pt: 53…58 °C Flash Pt: 98.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00179479 CAS Number: 251-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...