Thieno[3,2-b]pyridin-7-ol 95%

Supplier: Ambeed
Warning

A127964-250MG A127964-1G A127964-5G
103801-044EA 12 USD
103801-044 76782-144 77313-322 77313-324
Thieno[3,2-b]pyridin-7-ol 95%
Thieno[3,2-b]pyridin-7-ol

Precaución: for Research Use Only

Formula: C₇H₅NOS
Boiling Pt: 321.4 °C (760 mm Hg)
Melting Pt: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD01863636
CAS Number: 107818-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR