Thieno[2,3-d]pyrimidin-4-ylamine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Aminothieno[2,3-d]pyrimidine, Thieno[2,3-d]pyrimidin-4-amine

A181784-250MG A181784-1G
103807-646EA 66 USD
103807-646 103846-020 76784-602 77301-744
Thieno[2,3-d]pyrimidin-4-ylamine 97%
Thieno[2,3-d]pyrimidin-4-ylamine

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MW: 151.19 g/mol
Boiling Pt: 346.8 °C (760 mm Hg)
Melting Pt: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD01316189
CAS Number: 14080-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR