Thieno[2,3-d]pyrimidin-4(3H)-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Thieno[2,3-d]pyrimidin-4(1H)-one, 3,4-Dihydro-4-oxothieno[2,3-d]pyrimidine
A123468-100G
A123468-10G
A123468-25G
A123468-1G
A123468-5G
103800-408EA
8
USD
103800-408
77094-582
77094-584
77094-586
77297-944
77312-526
Thieno[2,3-d]pyrimidin-4(3H)-one 98%
Thieno[2,3-d]pyrimidin-4(3H)-one
Precaución: for Research Use Only
Formula:
C₆H₄N₂OS MW: 152.1738 g/mol Boiling Pt: 391.1 °C (760 mm Hg) Melting Pt: 262…265 °C Flash Pt: 190.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00706456 CAS Number: 14080-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...