Thieno[2,3-b]pyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A221827-100MG A221827-250MG
103818-948EA 51 USD
103818-948 103849-828 76786-010 76786-012
Thieno[2,3-b]pyridine-2-carboxylic acid 97%
Thieno[2,3-b]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂S
Melting Pt: 293…294 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371381
CAS Number: 59944-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR