Thiazole-5-carboxyaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 5-Thiazolecarboxaldehyde, 5-Formylthiazole

A236596-100G A236596-10G A236596-1G A236596-25G A236596-5G
103811-110EA 6 USD
103811-110 103850-884 76815-294 76815-296 77089-936 77089-938 77089-940
Thiazole-5-carboxyaldehyde 97%
Thiazole-5-carboxyaldehyde

Precaución: for Research Use Only

Formula: C₄H₃NOS
MW: 113.14 g/mol
Boiling Pt: 92…94 °C at 0,5 mmHg
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD02179516
CAS Number: 1003-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR