Thiazole-4-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Thiazole-4-carbaldehyde

A272289-100G A272289-10G A272289-250MG A272289-1G A272289-5G
103813-088EA 33 USD
103813-088 103852-998 76816-214 76816-216 76816-218 77092-788 77092-790
Thiazole-4-carboxaldehyde 97%
Thiazole-4-carboxaldehyde

Precaución: for Research Use Only

Formula: C₄H₃NOS
Boiling Pt: 222.4 °C (760 mm Hg)
Melting Pt: 60…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00626896
CAS Number: 3364-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR