2-Thiazolecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Thiazole-2-carboxylic acid

A126971-1G A126971-5G A126971-10G A126971-100G
103800-890EA 19 USD
103800-890 77313-134 77313-136 77549-508 77670-790
2-Thiazolecarboxylic acid 97%
2-Thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃NO₂S
MW: 129.14 g/mol
Boiling Pt: 310.6 °C (760 mm Hg)
Melting Pt: 91.6…93 °C
Flash Pt: 141.6 °C
Storage Temperature: Freezer
MDL Number: MFCD02094169
CAS Number: 14190-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR