5-Thiazolemethanol 97%

Supplier: Ambeed
Danger

Synonyms: Thiazol-5-ylmethanol, 5-(Hydroxymethyl)thiazole, Thiazole-5-methanol

A215012-1G A215012-5G A215012-100G A215012-10G A215012-500G
103849-152EA 7 USD
103849-152 76873-784 77087-678 77593-396 77671-916 77671-918
5-Thiazolemethanol 97%
5-Thiazolemethanol

Precaución: for Research Use Only

Formula: C₄H₅NOS
MW: 115.15 g/mol
Boiling Pt: 248.5 °C (760 mm Hg)
Flash Pt: 104.1 °C
Storage Temperature: Freezer
MDL Number: MFCD04115732
CAS Number: 38585-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR