5-(Aminomethyl)-1,3-thiazole hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Thiazol-5-ylmethanamine hydrochloride, (1,3-Thiazol-5-yl)methylamine hydrochloride

A161001-100MG A161001-250MG
103805-440EA 33 USD
103805-440 77011-196 77011-198
5-(Aminomethyl)-1,3-thiazole hydrochloride 97%
5-(Aminomethyl)-1,3-thiazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₆N₂S·ClH
MW: 150.63 g/mol
Melting Pt: 215…222 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738808
CAS Number: 131052-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR