Thiazol-2-ylmethanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A134697-250MG A134697-1G A134697-5G
103802-040EA 34 USD
103802-040 76811-442 77298-720 77298-722
Thiazol-2-ylmethanamine dihydrochloride 97%
Thiazol-2-ylmethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₈Cl₂N₂S
MW: 187.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09817422
CAS Number: 53332-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR