Benzo[b]thiophene 98%

Supplier: Ambeed
Danger

Synonyms: Thianaphthene, 1-Benzothiophene

A303896-25G A303896-10G A303896-500G A303896-5G
103835-564EA 6 USD
103835-564 76838-254 76875-678 76875-680 77305-890
Benzo[b]thiophene 98%
Benzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₆S
MW: 134.2 g/mol
Boiling Pt: 221…222 °C (760 mm Hg)
Melting Pt: 30…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00005864
CAS Number: 95-15-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR