N-(Trifluoroacetyl)glycine 97%

Supplier: Ambeed
Warning

Synonyms: (2,2,2-Trifluoroacetyl)glycine, Tfa-Gly-OH

A306534-100G A306534-10G A306534-1G A306534-25G A306534-5G
103814-474EA 9 USD
103814-474 103835-640 76787-856 76787-858 77305-934 77305-936 77305-938
N-(Trifluoroacetyl)glycine 97%
N-(Trifluoroacetyl)glycine

Precaución: for Research Use Only

Formula: C₄H₄F₃NO₃
MW: 171.075 g/mol
Boiling Pt: 291.6 °C (760 mm Hg)
Flash Pt: 130.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00029784
CAS Number: 383-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR