2,3,5-Triphenyltetrazolium chloride (TTC) 98%

Supplier: Ambeed
Danger

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

A244047-10G A244047-1G A244047-1MG A244047-25G A244047-5G
103851-390EA 17 USD
103851-390 77249-850 77249-852 77249-854 77304-302 77304-304
2,3,5-Triphenyltetrazolium chloride (TTC) 98%
2,3,5-Triphenyltetrazolium chloride (TTC)

Precaución: for Research Use Only

Formula: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Melting Pt: 235
Flash Pt: 91.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011963
CAS Number: 298-96-4
UN: 1325
ADR: 4.1 Ⅱ
Merck Index: 13,09815

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR