Tetra-n-butylammonium Di-tert-butylphosphate 95%

Supplier: Ambeed
Warning

A179473-10G A179473-250MG A179473-1G A179473-25G A179473-5G
103807-432EA 9 USD
103807-432 103845-780 76998-614 76998-616 77083-240 77083-242 77083-244
Tetra-n-butylammonium Di-tert-butylphosphate 95%
Tetra-n-butylammonium Di-tert-butylphosphate

Precaución: for Research Use Only

Formula: C₂₄H₅₄NO₄P
Storage Temperature: Ambient
MDL Number: MFCD12198885
CAS Number: 68695-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR