Tetrakis(triphenylphosphine)palladium(0) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
A211391-10G
A211391-25G
A211391-5G
103818-106EA
34
USD
103818-106
103848-800
76999-618
77087-206
77087-208
Tetrakis(triphenylphosphine)palladium(0) 98%
Tetrakis(triphenylphosphine)palladium(0)
Precaución: for Research Use Only
Formula:
C₇₂H₆₀P₄Pd MW: 1155.58 g/mol Melting Pt: 100…105 °C Flash Pt: 181.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...