Tetrakis(2,4-di-tert-butylphenyl) [1,1'-biphenyl]-4,4'-diylbis(phosphonite) 97%

Supplier: Ambeed
Warning

A127446-1G A127446-25G
103800-964EA 13 USD
103800-964 77298-212 77298-214
Tetrakis(2,4-di-tert-butylphenyl) [1,1'-biphenyl]-4,4'-diylbis(phosphonite) 97%
Tetrakis(2,4-di-tert-butylphenyl) [1,1'-biphenyl]-4,4'-diylbis(phosphonite)

Precaución: for Research Use Only

Formula: C₆₈H₉₂O₄P₂
MW: 1035.4 g/mol
Boiling Pt: 854.2 °C (760 mm Hg)
Melting Pt: 75…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799590
CAS Number: 38613-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR