Tetrahydroxyquinone 98+%

Supplier: Ambeed
Warning

Synonyms: Tetrahydroxy-1,4-Benzoquinone, 2,3,5,6-Tetrahydroxycyclohexa-2,5-diene-1,4-dione

A118395-100MG A118395-25MG
103799-048EA 23 USD
103799-048 76781-544 76781-546
Tetrahydroxyquinone 98+%
Tetrahydroxyquinone

Precaución: for Research Use Only

Formula: C₆H₄O₆
MW: 172.09 g/mol
Boiling Pt: 370.6 °C (760 mm Hg)
Melting Pt: 300 °C
Flash Pt: 192.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001597
CAS Number: 319-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR