Tetrahydropyran-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Tetrahydropyran-4-yl-carboxylic acid

A113172-100G A113172-10G A113172-25G A113172-500G
103824-016EA 7 USD
103824-016 77160-858 77160-860 77160-862 77160-864
Tetrahydropyran-4-carboxylic acid 97%
Tetrahydropyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 264.5 °C (760 mm Hg)
Melting Pt: 87…89 °C
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00031016
CAS Number: 5337-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR