Tetrahydropyran-4-methanol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Hydroxymethyl)tetrahydropyran

A215491-25G A215491-5G A215491-100G A215491-10G A215491-500G A215491-1G
103818-494EA 5 USD
103818-494 103849-202 77087-738 77087-740 77152-600 77152-602 77152-604 77303-412
Tetrahydropyran-4-methanol 98%
Tetrahydropyran-4-methanol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 218.9 °C (760 mm Hg)
Flash Pt: 97.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00457804
CAS Number: 14774-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR