Tetrahydropentalene-2,5(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A138352-10G A138352-250MG A138352-1G A138352-25G
103802-600EA 32 USD
103802-600 76782-788 76782-790 77315-618 77315-620
Tetrahydropentalene-2,5(1H,3H)-dione 95%
Tetrahydropentalene-2,5(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
Boiling Pt: 268.3 °C (760 mm Hg)
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00071658
CAS Number: 74513-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR