Tetrahydrofuran-3-amine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Aminotetrahydrofuran hydrochloride
A213685-250MG
A213685-25G
A213685-1G
103818-314EA
11
USD
103818-314
103849-022
76785-754
76785-756
77087-532
Tetrahydrofuran-3-amine hydrochloride 95%
Tetrahydrofuran-3-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₀ClNO MW: 123.5800 g/mol Boiling Pt: 173.5 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD06738925 CAS Number: 204512-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...