(±)-Tetrahydro-2-furoic acid 97%

Supplier: Ambeed
Danger

A444879-25G A444879-100G A444879-10G A444879-5G
103810-346EA 176 USD
103810-346 103838-788 76842-568 77004-276 77004-278 77307-334
(±)-Tetrahydro-2-furoic acid 97%
(±)-Tetrahydro-2-furoic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
Boiling Pt: 243.2 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR