Tetrahydrofolic acid 67%

Supplier: Ambeed
Warning

Synonyms: 5,6,7,8-Tetrahydropteroyl-L-glutamic acid, THF, (2S)-2-(4-(((2-Amino-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid

A276659-100MG A276659-250MG
103813-316EA 716 USD
103813-316 76787-298 76787-300
Tetrahydrofolic acid 67%
Tetrahydrofolic acid

Precaución: for Research Use Only

Formula: C₁₉H₂₃N₇O₆
MW: 445.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135583
CAS Number: 135-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR