Tetrahydrocyclopenta[c]pyrrole-1,3(2H,3aH)-dione 97%

Supplier: Ambeed
Warning

A191091-100G A191091-500G A191091-5G A191091-1000G A191091-25G A191091-10G
103809-252EA 291 USD
103809-252 76690-892 76690-894 76690-896 77084-778 77084-780 77625-918
Tetrahydrocyclopenta[c]pyrrole-1,3(2H,3aH)-dione 97%
Tetrahydrocyclopenta[c]pyrrole-1,3(2H,3aH)-dione

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 322.2 °C (760 mm Hg)
Melting Pt: 168 °C
Flash Pt: 162.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06411094
CAS Number: 5763-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR