Tetrahydro-2H-thiopyran-4-ol 97%

Supplier: Ambeed
Danger

A125222-100MG A125222-250MG A125222-1G
103800-644EA 26 USD
103800-644 76781-960 76781-962 77312-792
Tetrahydro-2H-thiopyran-4-ol 97%
Tetrahydro-2H-thiopyran-4-ol

Precaución: for Research Use Only

Formula: C₅H₁₀OS
MW: 118.2 g/mol
Boiling Pt: 218.8 °C (760 mm Hg)
Melting Pt: 47…51 °C (760 mm Hg)
Flash Pt: 107.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02942489
CAS Number: 29683-23-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR