Tetrahydro-2H-pyran-4-yl methanesulfonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Tetrahydropyranyl methanesulphonate
A254903-25G
A254903-5G
A254903-1G
A254903-100G
A254903-10G
A254903-500G
103812-176EA
9
USD
103812-176
103852-030
76786-882
77091-488
77304-644
77550-620
77568-468
77568-470
Tetrahydro-2H-pyran-4-yl methanesulfonate 97%
Tetrahydro-2H-pyran-4-yl methanesulfonate
Precaución: For research use only.
Formula:
C₆H₁₂O₄S MW: 180.218 g/mol Boiling Pt: 329.1 °C (760 mm Hg) Melting Pt: 49…50 °C Flash Pt: 152.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD11505063 CAS Number: 134419-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...