Tetrahydro-2H-pyran-4-carbonyl chloride 97%
Supplier: Ambeed
|
Danger
|
A342713-10G
A342713-1G
A342713-25G
A342713-5G
103825-868EA
15
USD
103825-868
103836-842
76788-302
76839-886
76839-888
76839-890
Tetrahydro-2H-pyran-4-carbonyl chloride 97%
Tetrahydro-2H-pyran-4-carbonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₉ClO₂ Boiling Pt: 195.6 °C (760 mm Hg) Flash Pt: 71.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200863 CAS Number: 40191-32-0 UN: 3265 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...