4-Aminotetrahydropyran 97%

Supplier: Ambeed
Danger

Synonyms: Tetrahydro-2H-pyran-4-amine

A348036-25G A348036-5G A348036-10G
103825-984EA 5 USD
103825-984 103836-970 76840-044 76840-046 76989-564
4-Aminotetrahydropyran 97%
4-Aminotetrahydropyran

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.1469 g/mol
Boiling Pt: 151.4 °C (760 mm Hg)
Flash Pt: 54.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179436
CAS Number: 38041-19-9
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR