Tetrahydro-2H-pyran-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A256185-1G A256185-25G A256185-5G A256185-10G A256185-250MG
103812-244EA 34 USD
103812-244 103852-108 77091-588 77091-590 77091-592 77153-344 77153-346
Tetrahydro-2H-pyran-3-carboxylic acid 97%
Tetrahydro-2H-pyran-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 264.5 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114349
CAS Number: 873397-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR