δ-Valerolactone 98% may contain polymer

Supplier: Ambeed
Danger

Synonyms: Pentan-5-olide, Tetrahydro-2H-pyran-2-one, 5-Hydroxypentanoic acid lactone, δ-Valerolactone

A166595-10G A166595-25G A166595-100G A166595-500G
103808-972EA 104 USD
103808-972 76690-074 77320-614 77550-064 77593-006
δ-Valerolactone 98% may contain polymer
δ-Valerolactone

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 218 °C (760 mm Hg)
Melting Pt: …13 °C
Flash Pt: 81 °C
Storage Temperature: Freezer
MDL Number: MFCD00006645
CAS Number: 542-28-9
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR