Tetrahydropyran-2-carboxylic acid 97%

Supplier: Ambeed
Danger

A199468-10G A199468-1G A199468-5G A199468-25G
103817-108EA 12 USD
103817-108 103847-732 76785-260 77085-774 77085-776 77871-994
Tetrahydropyran-2-carboxylic acid 97%
Tetrahydropyran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
Boiling Pt: 264.5 °C (760 mm Hg)
Flash Pt: 114.039 °C
Storage Temperature: Ambient
MDL Number: MFCD07779239
CAS Number: 51673-83-7
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR