Tetrafluoroterephthalonitrile 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Perfluoroterephthalonitrile
A140249-100G
A140249-500G
A140249-25G
A140249-5G
77027-480EA
130.5
USD
77027-480
77027-482
77299-122
77299-124
Tetrafluoroterephthalonitrile 98%
Tetrafluoroterephthalonitrile
Precaución: for Research Use Only
Formula:
C₈F₄N₂ MW: 200.1 g/mol Boiling Pt: 243.3 °C (760 mm Hg) Melting Pt: 197…199 °C Flash Pt: 100.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001776 CAS Number: 1835-49-0 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...