Tetraethylammonium toluene-4-sulfonate 98%

Supplier: Ambeed
Danger

Synonyms: Tetraethylammonium p-toluenesulphonate, Tetraethylammonium p-Toluenesulphonate

A551535-10G A551535-25G A551535-5G
103839-824EA 6 USD
103839-824 76790-254 76843-894 76843-896
Tetraethylammonium toluene-4-sulfonate 98%
Tetraethylammonium toluene-4-sulphonate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₃S
MW: 301.4500 g/mol
Melting Pt: 108…114 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD00011831
CAS Number: 733-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR