Tetrabutylammonium hydrogen sulfate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate
A133607-100G
A133607-500G
A133607-1000G
A133607-25G
103808-470EA
47
USD
103808-470
76782-496
76782-498
77298-638
77298-640
Tetrabutylammonium hydrogen sulfate 98%
Tetrabutylammonium hydrogen sulphate
Precaución: for Research Use Only
Formula:
(C₄H₉)₄NHSO₄ MW: 339.54 g/mol Melting Pt: 171…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011637 CAS Number: 32503-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...