Tetrabutylammonium hexafluorophosphate 97%

Supplier: Ambeed
Warning

A232855-100G A232855-10G A232855-500G A232855-1000G A232855-25G A232855-5G
103809-640EA 223 USD
103809-640 103850-614 76786-308 76786-310 76786-312 77089-550 77089-552 77303-948
Tetrabutylammonium hexafluorophosphate 97%
Tetrabutylammonium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₆H₃₆F₆NP
Melting Pt: 242…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011748
CAS Number: 3109-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR