Tetrabutylammonium fluoride trihydrate 95%

Supplier: Ambeed
Danger

A213236-100G A213236-10G A213236-500G A213236-25G
103809-482EA 92 USD
103809-482 76785-740 76785-742 76785-744 77087-470 77087-472
Tetrabutylammonium fluoride trihydrate 95%
Tetrabutylammonium fluoride trihydrate

Precaución: for Research Use Only

Formula: C₁₆H₃₆FN·3H₂O
MW: 315.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149981
CAS Number: 87749-50-6
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR