Tetrabutylammonium bromide 98%

Supplier: Ambeed
Warning

A417768-1000G A417768-100G A417768-500G
103810-310EA 29 USD
103810-310 76841-978 77115-906 77115-908
Tetrabutylammonium bromide 98%
Tetrabutylammonium bromide

Precaución: for Research Use Only

Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00011633
CAS Number: 1643-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR