tert-Butyl trichloroacetimidate 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 2,2,2-trichloroacetimidate, t-Butyl trichioroacetimidate, t-butyl-2,2,2-trichloroacetimidate

A817163-10G A817163-1G A817163-5G
103841-110EA 8 USD
103841-110 76791-694 76845-350 77308-374
tert-Butyl trichloroacetimidate 98%
tert-Butyl trichloroacetimidate

Precaución: for Research Use Only

Formula: C₆H₁₀Cl₃NO
MW: 218.502 g/mol
Boiling Pt: 183.3 °C (760 mm Hg)
Melting Pt: 21 °C
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077410
CAS Number: 98946-18-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR