tert-Butyl thiophen-2-ylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminothiophene, N-BOC protected

A175299-10G A175299-250MG A175299-1G A175299-5G
103807-030EA 11 USD
103807-030 103845-334 76784-410 76784-412 77082-676 77082-678
tert-Butyl thiophen-2-ylcarbamate 97%
tert-Butyl thiophen-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MW: 199.2700 g/mol
Boiling Pt: 238.1 °C (760 mm Hg)
Melting Pt: 151 °C
Flash Pt: 97.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02677742
CAS Number: 56267-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR