tert-Butyl propiolate 95%

Supplier: Ambeed
Danger

A193947-10G A193947-1G A193947-25G A193947-5G
103816-602EA 13 USD
103816-602 103847-210 76785-048 77085-114 77085-116 77085-118
tert-Butyl propiolate 95%
tert-Butyl propiolate

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
Boiling Pt: 159.6 °C (760 mm Hg)
Melting Pt: 18…20 °C
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060100
CAS Number: 13831-03-3
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR